Learn More
Medchemexpress LLC PBX-7011 TFA | 98.6% | 561.46 | 5 MG
SDP
Catalog No. 5000391370
Encompass_Preferred
PBX-7011 TFA is a derivative of camptothecin. It inhibits expressions of the cancer-related survival genes DDX5, Survivin, Mcl-1, and XIAP in FaDu cells, degrades DDX5 proteins, and exhibits anticancer activity.
  • Inhibits expressions of cancer-related survival genes DDX5, Survivin, Mcl-1, and XIAP in FaDu cells.
  • Degrades DDX5 proteins.
  • Exhibits anticancer activity.
Catalog No. 50-003-91370 Supplier Medchemexpress LLC Supplier No. HY160438B5MG
PBX-7011 TFA is a derivative of camptothecin. It inhibits expressions of the cancer-related survival genes DDX5, Survivin, Mcl-1, and XIAP in FaDu cells, degrades DDX5 proteins, and exhibits anticancer activity.
  • Inhibits expressions of cancer-related survival genes DDX5, Survivin, Mcl-1, and XIAP in FaDu cells.
  • Degrades DDX5 proteins.
  • Exhibits anticancer activity.