Learn More
Aviva Systems Biology Corp PBXIP1 Antibody - N-terminal region (ARP32948_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32948_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PBXIP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PBXIP1. Alternate names: HPIPApplicationWB. Species reactivity: Cow, Human, Mouse, Rat
Catalog No. 50-014-54826