Learn More
Biosurface Technologies Corp Dual Channel Transmission Flow Cell w/injection ports (polycarbonate flow channel; platform stage mount), for Growing and Imaging of Biofilms.
Catalog No. NC3662474
Encompass
"The models FC 284 and 285 flow cell design is based on the FC 81 but accommodates two growth channels in the same footprint as a 96-well microtiter plate. This format allows biofilms to be grown side by side for duplicate or “control-treatment” experiments. Biofilms growing in each channel can be easily compared microscopically. The body of the flow cell consists of a polycarbonate or anodized aluminum flow channel. Glass viewing ports allow transmitted, reflected, fluorescent, and confocal microscopy of biofilm growth in the flow channels. All units are autoclavable and re-useable. The flow cell dimensions allow for easy mounting in inverted microscope stages designed to hold 96-well microtiter plates.The viewing windows consist of a no. 2, 24x60 mm cover slip. The opposing viewing window consists of a standard 75 x 25 x 1 mm glass microscope slide. The glass viewing windows are held in place by an aluminum cover plate.
Catalog No. NC3662474 Supplier Biosurface Technologies Corp Supplier No. FC 284PC
"The models FC 284 and 285 flow cell design is based on the FC 81 but accommodates two growth channels in the same footprint as a 96-well microtiter plate. This format allows biofilms to be grown side by side for duplicate or “control-treatment” experiments. Biofilms growing in each channel can be easily compared microscopically. The body of the flow cell consists of a polycarbonate or anodized aluminum flow channel. Glass viewing ports allow transmitted, reflected, fluorescent, and confocal microscopy of biofilm growth in the flow channels. All units are autoclavable and re-useable. The flow cell dimensions allow for easy mounting in inverted microscope stages designed to hold 96-well microtiter plates.The viewing windows consist of a no. 2, 24x60 mm cover slip. The opposing viewing window consists of a standard 75 x 25 x 1 mm glass microscope slide. The glass viewing windows are held in place by an aluminum cover plate.