Learn More
Quip Laboratories Inc PC-500, 5 Gallon
SDP
Catalog No. NC9070811
Encompass
PC-500 is a powerful liquid blend of chelating agents, sequesterants and alkaline builders, designed to tackle a wide range of tough cleaning problems. Need to remove deposits from glassware? Look no further than PC-500. Safe on metals found in most glassware washers. Cleans and rinses glassware free of deposits without chlorine. Effective in removing certain types of laboratory markers from glassware.
This item is not returnable. View return policy
Catalog No. NC9070811 Supplier Quip Laboratories Inc Supplier No. PC5005GAL
PC-500 is a powerful liquid blend of chelating agents, sequesterants and alkaline builders, designed to tackle a wide range of tough cleaning problems. Need to remove deposits from glassware? Look no further than PC-500. Safe on metals found in most glassware washers. Cleans and rinses glassware free of deposits without chlorine. Effective in removing certain types of laboratory markers from glassware.