Learn More
ATCC PC 61 5.3; Hybridoma; Rat (Rattus norvegicus)/Mouse (Mus muscullus)
Catalog No. 502383278
Encompass
PC 61 5.3 [PC 61, PC 61.5.3] is a B lymphocyte cell that was isolated from the spleen of a mouse with leukemia and can be used in immunology and immune system disorder research.
Catalog No. 50-238-3278 Supplier ATCC Supplier No. TIB222
PC 61 5.3 [PC 61, PC 61.5.3] is a B lymphocyte cell that was isolated from the spleen of a mouse with leukemia and can be used in immunology and immune system disorder research.