Learn More
Electron Microscopy Sciences PC-620 Ultrasonic Cleaner with Heat 230/240 V
SDP
Catalog No. 50930728
Encompass_Preferred
Ultrasonic cleaner ideal for cleaning pipettes, lab glassware, and other items requiring general cleaning. Capacity: 2.75 gallons, tank dimensions: 19.5"L x 5 3/4"W x 6"D, overall dimensions: 20.5"L x 61 1/4"W x 10.5"W.
Catalog No. 50-930-728 Supplier Electron Microscopy Sciences Supplier No. 71046E2
Ultrasonic cleaner ideal for cleaning pipettes, lab glassware, and other items requiring general cleaning. Capacity: 2.75 gallons, tank dimensions: 19.5"L x 5 3/4"W x 6"D, overall dimensions: 20.5"L x 61 1/4"W x 10.5"W.