Learn More
Medchemexpress LLC PC azido-PEG3-NHS carbonate ester | 1802907-96-5 | MFCD29052189 | 98.7% | 640.60 | C26H36N6O13 | 10 MG

Supplier: Medchemexpress LLC HYW19093710MG
PC Azido-PEG3-NHS carbonate ester is a photocleavable linker used to attach an azido-terminated PEG3 spacer to primary amines on proteins, amine-modified oligonucleotides, and other amine-containing molecules via an NHS carbonate ester, enabling bioorthogonal click chemistry and light-triggered release.
- Photocleavable linker for light-triggered release
- Labels primary amines on proteins and oligonucleotides
- Installs azido-terminated PEG3 spacer via NHS carbonate chemistry
- Enables copper-catalyzed azide-alkyne click reactions
- High purity suitable for bioconjugation (≈98.7%)
- Soluble in DMSO at ≥116.67 mg/mL
- Store at -20 °C in pure form for long-term stability
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.