Learn More
Medchemexpress LLC PC azido-PEG3-NHS carbonate ester | 1802907-96-5 | MFCD29052189 | 98.7% | 640.60 | C26H36N6O13 | 100 MG
SDP
Catalog No. 5000281621
Encompass_Preferred
PC Azido-PEG3-NHS carbonate ester is a photocleavable linker reagent that selectively labels primary amines on proteins, amine-modified oligonucleotides, and other amine-containing molecules. It contains an azide for bioorthogonal click chemistry and an N-hydroxysuccinimide (NHS) carbonate group for efficient coupling; the photocleavable moiety enables light-triggered release of attached cargos. Supplied at high purity for research use.
  • Photocleavable linker enabling light-triggered release
  • Reacts with primary amines via NHS carbonate chemistry
  • Includes an azide for bioorthogonal click reactions
  • High purity suitable for research applications
  • Offered in multiple pack sizes, e.g., 100 mg
Catalog No. 50-002-81621 Supplier Medchemexpress LLC Supplier No. HYW190937100MG
PC Azido-PEG3-NHS carbonate ester is a photocleavable linker reagent that selectively labels primary amines on proteins, amine-modified oligonucleotides, and other amine-containing molecules. It contains an azide for bioorthogonal click chemistry and an N-hydroxysuccinimide (NHS) carbonate group for efficient coupling; the photocleavable moiety enables light-triggered release of attached cargos. Supplied at high purity for research use.
  • Photocleavable linker enabling light-triggered release
  • Reacts with primary amines via NHS carbonate chemistry
  • Includes an azide for bioorthogonal click reactions
  • High purity suitable for research applications
  • Offered in multiple pack sizes, e.g., 100 mg