Learn More
Medchemexpress LLC PC azido-PEG3-NHS carbonate ester | 1802907-96-5 | MFCD29052189 | 98.7% | 640.60 | C26H36N6O13 | 25 MG
SDP
Catalog No. 5000283165
Encompass_Preferred
PC Azido-PEG3-NHS carbonate ester is a photocleavable carbonate ester linker used to label primary amines on proteins, amine-modified oligonucleotides, and other amine-containing molecules. It contains an azido group for bioorthogonal conjugation and a PEG3 spacer to enhance solubility and reduce steric hindrance. Supplied as an oil for research use, it is stored frozen and is soluble in DMSO.
  • Photocleavable carbonate chemistry enables light-triggered release.
  • Azido functional group supports bioorthogonal click reactions.
  • PEG3 spacer improves solubility and conjugation efficiency.
  • High purity suitable for research applications.
  • Supplied as an oil with recommended cold storage.
  • Soluble in DMSO; use freshly opened DMSO to ensure solubility.
Catalog No. 50-002-83165 Supplier Medchemexpress LLC Supplier No. HYW19093725MG
PC Azido-PEG3-NHS carbonate ester is a photocleavable carbonate ester linker used to label primary amines on proteins, amine-modified oligonucleotides, and other amine-containing molecules. It contains an azido group for bioorthogonal conjugation and a PEG3 spacer to enhance solubility and reduce steric hindrance. Supplied as an oil for research use, it is stored frozen and is soluble in DMSO.
  • Photocleavable carbonate chemistry enables light-triggered release.
  • Azido functional group supports bioorthogonal click reactions.
  • PEG3 spacer improves solubility and conjugation efficiency.
  • High purity suitable for research applications.
  • Supplied as an oil with recommended cold storage.
  • Soluble in DMSO; use freshly opened DMSO to ensure solubility.