Learn More
Medchemexpress LLC PC azido-PEG3-NHS carbonate ester | 1802907-96-5 | MFCD29052189 | 98.7% | 640.60 g·mol⁻¹ | C26H36N6O13 | 5 MG
SDP
Catalog No. 5000280773
Encompass_Preferred
PC Azido-PEG3-NHS carbonate ester is a photocleavable, heterobifunctional linker designed for covalent labeling of primary amines and for introducing an azide handle for click chemistry. The reagent couples to amine groups via an NHS carbonate, and the photocleavable moiety enables light-triggered release of the attached cargo; supplied as a dry research reagent with recommended cold storage.
Catalog No. 50-002-80773 Supplier Medchemexpress LLC Supplier No. HYW1909375MG
PC Azido-PEG3-NHS carbonate ester is a photocleavable, heterobifunctional linker designed for covalent labeling of primary amines and for introducing an azide handle for click chemistry. The reagent couples to amine groups via an NHS carbonate, and the photocleavable moiety enables light-triggered release of the attached cargo; supplied as a dry research reagent with recommended cold storage.