Learn More
Medchemexpress LLC Pc biotin-PEG3-azide | 1937270-46-6 | 97.8% | 825.93 g/mol | C35H55N9O12S | 10 MG
SDP
Catalog No. 5000202276
Encompass_Preferred
PC Biotin-PEG3-azide is a cleavable three-unit polyethylene glycol (PEG3) biotinylated azide linker used for bioconjugation and the synthesis of antibody-drug conjugates. It contains an azide functional group for click chemistry and reacts in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Cleavable three-unit PEG linker.
  • Contains an azide functional group for click chemistry.
  • Suits copper-catalyzed and strain-promoted azide-alkyne cycloadditions.
  • High research-grade purity.
  • Available in small-scale quantities for conjugation workflows.
Catalog No. 50-002-02276 Supplier Medchemexpress LLC Supplier No. HY14013210MG
PC Biotin-PEG3-azide is a cleavable three-unit polyethylene glycol (PEG3) biotinylated azide linker used for bioconjugation and the synthesis of antibody-drug conjugates. It contains an azide functional group for click chemistry and reacts in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Cleavable three-unit PEG linker.
  • Contains an azide functional group for click chemistry.
  • Suits copper-catalyzed and strain-promoted azide-alkyne cycloadditions.
  • High research-grade purity.
  • Available in small-scale quantities for conjugation workflows.