Learn More
Medchemexpress LLC Pc biotin-peg3-nhs ester | 2353409-93-3 | 95.0% | 840.89 g/mol | C36H52N6O15S | 5 MG
SDP
Catalog No. 5000202622
Encompass_Preferred
PC Biotin-PEG3-NHS ester is a cleavable three-unit polyethylene glycol (PEG3) linker bearing a biotin tag and an N-hydroxysuccinimide (NHS) ester for coupling to primary amines, used in linker and biotinylation chemistries.
  • Cleavable PEG3 linker suitable for conjugation chemistries.
  • Contains an NHS ester for efficient reaction with primary amines.
  • Biotin tag enables affinity capture and detection.
  • Supplied in small, research-scale quantities for synthetic workflows.
  • Stable when stored as recommended; purity suitable for research use.
Catalog No. 50-002-02622 Supplier Medchemexpress LLC Supplier No. HY1401345MG
PC Biotin-PEG3-NHS ester is a cleavable three-unit polyethylene glycol (PEG3) linker bearing a biotin tag and an N-hydroxysuccinimide (NHS) ester for coupling to primary amines, used in linker and biotinylation chemistries.
  • Cleavable PEG3 linker suitable for conjugation chemistries.
  • Contains an NHS ester for efficient reaction with primary amines.
  • Biotin tag enables affinity capture and detection.
  • Supplied in small, research-scale quantities for synthetic workflows.
  • Stable when stored as recommended; purity suitable for research use.