Learn More
Medchemexpress LLC PC-biotin-PEG4-NHS carbonate | 2055198-03-1 | 97.0% | C35H50N6O14S | 10MG
SDP
Catalog No. 5000260399
Encompass_Preferred
PC-Biotin-PEG4-NHS carbonate is a PEG-based photocleavable biotinylation reagent bearing an NHS carbonate amine-reactive group. It is designed for use as a PROTAC linker or to introduce a light-cleavable biotin tag for biochemical conjugation, affinity capture, and downstream light-mediated release of labeled molecules.
  • Photocleavable biotin enables light-controlled release of captured molecules.
  • PEG4 spacer improves solubility and reduces steric hindrance in conjugates.
  • NHS carbonate reacts with primary amines to form stable carbamate linkages.
  • High reported purity supports use in synthesis and analytical workflows.
  • Powder storage at -20°C and cold-solvent storage extend compound stability.
Catalog No. 50-002-60399 Supplier Medchemexpress LLC Supplier No. HY14013510MG
PC-Biotin-PEG4-NHS carbonate is a PEG-based photocleavable biotinylation reagent bearing an NHS carbonate amine-reactive group. It is designed for use as a PROTAC linker or to introduce a light-cleavable biotin tag for biochemical conjugation, affinity capture, and downstream light-mediated release of labeled molecules.
  • Photocleavable biotin enables light-controlled release of captured molecules.
  • PEG4 spacer improves solubility and reduces steric hindrance in conjugates.
  • NHS carbonate reacts with primary amines to form stable carbamate linkages.
  • High reported purity supports use in synthesis and analytical workflows.
  • Powder storage at -20°C and cold-solvent storage extend compound stability.