Learn More
Medchemexpress LLC PC-biotin-PEG4-NHS carbonate | 2055198-03-1 | MFCD20655073 | 97.0% | 810.87 | C35H50N6O14S | 5 MG
SDP
Catalog No. 5000222350
Encompass_Preferred
PC-Biotin-PEG4-NHS carbonate is a photocleavable PEG4-based biotinylation reagent with an NHS carbonate moiety that reacts with primary amines to form carbamate linkages. It is designed for affinity capture and light-controlled release of biomolecules, and can be used as a linker in chemical biology applications such as PROTAC synthesis.
  • Photocleavable biotin tag enables light-controlled release of captured molecules.
  • NHS carbonate reacts selectively with primary amines to form stable carbamate bonds.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • Suitable for affinity capture, labeling, and linker synthesis in chemical biology.
  • Compatible with near-UV illumination for efficient photorelease.
Catalog No. 50-002-22350 Supplier Medchemexpress LLC Supplier No. HY1401355MG
PC-Biotin-PEG4-NHS carbonate is a photocleavable PEG4-based biotinylation reagent with an NHS carbonate moiety that reacts with primary amines to form carbamate linkages. It is designed for affinity capture and light-controlled release of biomolecules, and can be used as a linker in chemical biology applications such as PROTAC synthesis.
  • Photocleavable biotin tag enables light-controlled release of captured molecules.
  • NHS carbonate reacts selectively with primary amines to form stable carbamate bonds.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • Suitable for affinity capture, labeling, and linker synthesis in chemical biology.
  • Compatible with near-UV illumination for efficient photorelease.