Learn More
Foxx Life Sciences PC Carboy 2.5L 83B Neck No Spigot 1/EA
Catalog No. 5000573859
Encompass_Preferred
EZBio PC Carboys are sterile fluid containers designed for high-performance bioprocessing and pharmaceutical-grade applications The carboy comes in a distinctive rectangular shape that saves valuable bench space and features a wide-mouth design for easy filling and cleaning The carboy body is ergonomically designed with large top handles and molded hand grips in the base allowing the user to easily hold and manipulate the instrument during filling lifting and pouring These cleanroom-compatible carboys support critical fluid handling in controlled environments br Br Features Br br Available in 2 5L 5L 10L and 20L sizes (Sterile and Non Sterile) br Molded from USP Class VI resins br FDA 21 CFR 177 compliant materials Br Clear body for easy visual inspection Br Graduation marks in liters Br Leakproof VersaCap 83B compatibility br Br Autoclavable Br Manufactured in a Class 7 cleanroom
Catalog No. 50-005-73859 Supplier Foxx Life Sciences Supplier No. 5185C51FLS
EZBio PC Carboys are sterile fluid containers designed for high-performance bioprocessing and pharmaceutical-grade applications The carboy comes in a distinctive rectangular shape that saves valuable bench space and features a wide-mouth design for easy filling and cleaning The carboy body is ergonomically designed with large top handles and molded hand grips in the base allowing the user to easily hold and manipulate the instrument during filling lifting and pouring These cleanroom-compatible carboys support critical fluid handling in controlled environments br Br Features Br br Available in 2 5L 5L 10L and 20L sizes (Sterile and Non Sterile) br Molded from USP Class VI resins br FDA 21 CFR 177 compliant materials Br Clear body for easy visual inspection Br Graduation marks in liters Br Leakproof VersaCap 83B compatibility br Br Autoclavable Br Manufactured in a Class 7 cleanroom