Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ PC Carboy

Catalog No. 14388180 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14-388-180 10 L
14-388-181 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-388-180
Add to Cart
Add to Cart

Large-volume media and culture preparation, particularly when visual inspection is required.; Autoclavable; Closure Material: Polypropylene; Color: Clear

  • Conduct large-volume media and culture preparation with these 10 L FISHERBRAND PC, Carboy, particularly when visual inspection is required.
  • This polycarbonate Clearboy is durable, lighter and safer than glass, autoclavable for sterile applications and ideal for refrigerated or frozen storage of aqueous solutions such as sterile water, aqueous reagents and media.
  • Features:
  • Available in 10 and 20 L sizes for a variety of biopharmaceutical applications
  • Graduated in liters and gallons
  • Molded of transparent, durable polycarbonate
  • Customization options available for fluid transfer applications
  • Autoclavable, graduated, leakproof
  • Includes: Polypropylene closure and TPE gasket
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (Metric) 10 L
Color Natural (Translucent)
Disposable Reusable
Material Polycarbonate
Mouth Style Wide
Product Type Carboy
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.