Learn More
Medchemexpress LLC PC DBCO-PEG4-NHS ester | 2055025-02-8 | 96.9% | C47H55N5O16 | 10MG
SDP
Catalog No. 5000261613
Encompass_Preferred
PC DBCO-PEG4-NHS ester is a photocleavable PEG-based linker containing a dibenzocyclooctyne (DBCO) moiety and an N-hydroxysuccinimide (NHS) carbonate reactive group. It is designed for bioconjugation workflows, enabling coupling to primary amines and strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-functionalized partners, with the option of light-triggered cleavage when required.
  • Photocleavable linker enabling light-triggered cleavage.
  • DBCO moiety for strain-promoted alkyne-azide cycloaddition (SPAAC).
  • NHS carbonate group for efficient coupling to primary amines.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • Suitable for bioconjugation workflows and PROTAC synthesis.
  • Supplied as a solid, white to light yellow, with high listed purity.
Catalog No. 50-002-61613 Supplier Medchemexpress LLC Supplier No. HY13875610MG
PC DBCO-PEG4-NHS ester is a photocleavable PEG-based linker containing a dibenzocyclooctyne (DBCO) moiety and an N-hydroxysuccinimide (NHS) carbonate reactive group. It is designed for bioconjugation workflows, enabling coupling to primary amines and strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-functionalized partners, with the option of light-triggered cleavage when required.
  • Photocleavable linker enabling light-triggered cleavage.
  • DBCO moiety for strain-promoted alkyne-azide cycloaddition (SPAAC).
  • NHS carbonate group for efficient coupling to primary amines.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • Suitable for bioconjugation workflows and PROTAC synthesis.
  • Supplied as a solid, white to light yellow, with high listed purity.