Learn More
Foxx Life Sciences EZBio 250mL PC Media Bottles, Sterile, Autoclavable Square Storage Bottles with 38-430mm White Closed VersaCap Closures, 24/PK
GREENER_CHOICE
Catalog No. 501613238
Encompass_Preferred
EZBio Polycarbonate Sterile Media Bottles are perfect for storage of sterile reagents and media, and can be used with EZBio Single-Use Cap Assemblies for aseptic transfer and sampling applications. Bottles withstand temperatures down to -125C for freezing applications, and can be autoclaved at 121C for 20 minutes up to ten times. Gamma-irradiated. Assembled in a Class 10,000 Cleanroom. Packaged in double-bagged trays. 100% USP Class VI, ADCF-free materials. VersaCaps with vented membranes or molded hose barbs available separately.
Catalog No. 50-161-3238 Supplier Foxx Life Sciences Supplier No. 1383211FLS
EZBio Polycarbonate Sterile Media Bottles are perfect for storage of sterile reagents and media, and can be used with EZBio Single-Use Cap Assemblies for aseptic transfer and sampling applications. Bottles withstand temperatures down to -125C for freezing applications, and can be autoclaved at 121C for 20 minutes up to ten times. Gamma-irradiated. Assembled in a Class 10,000 Cleanroom. Packaged in double-bagged trays. 100% USP Class VI, ADCF-free materials. VersaCaps with vented membranes or molded hose barbs available separately.