Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SI Analytics™ PC Keyboard

Catalog No. 15177888 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
For Use With (Equipment):
TitroLine 5000, TitroLine 7000 , TitroLine 7500 KF/7500 KF Trace, TitroLine 7750 and TITRONIC 300/500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
15-177-888 TitroLine 5000, TitroLine 7000 , TitroLine 7500 KF/7500 KF Trace, TitroLine 7750 and TITRONIC 300/500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-177-888 Supplier SI Analytics™ Supplier No. 285220410
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Autotitrators and Piston Burettes

TRUSTED_SUSTAINABILITY

Specifications

Connector Types USB
Type USB Keyboard
Height (English) 1 in.
Length (English) 4 in.
Weight (English) 1 lb.
Width (English) 4 in.
For Use With (Equipment) TitroLine 5000, TitroLine 7000 , TitroLine 7500 KF/7500 KF Trace, TitroLine 7750 and TITRONIC 300/500
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.