Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PC Mal-NHS carbonate ester | 1408057-91-9 | 95.0% | 562.48 | C24H26N4O12 | 5 MG
SDP

Catalog No. 5000202620
Encompass_Preferred

PC Mal-NHS carbonate ester is a photocleavable, cleavable antibody-drug conjugate (ADC) linker that contains maleimide and NHS carbonate functional groups; it enables conjugation to thiols and amines and controlled payload release under photocleavage conditions. Supplied for research use in small milligram quantities.

  • Cleavable photocleavable linker for controlled payload release.
  • Maleimide group reacts with thiols for thiol-specific conjugation.
  • NHS carbonate group reacts with amines for versatile payload attachment.
  • High purity (95.0%), white to off-white solid.
  • Soluble in DMSO (100 mg/mL; may require ultrasonic treatment).
  • Storage: powder -20 °C; in solvent -80 °C (6 months) or -20 °C (1 month).

Catalog No. 50-002-02620 Supplier Medchemexpress LLC Supplier No. HY1401405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PC Mal-NHS carbonate ester is a photocleavable, cleavable antibody-drug conjugate (ADC) linker that contains maleimide and NHS carbonate functional groups; it enables conjugation to thiols and amines and controlled payload release under photocleavage conditions. Supplied for research use in small milligram quantities.

  • Cleavable photocleavable linker for controlled payload release.
  • Maleimide group reacts with thiols for thiol-specific conjugation.
  • NHS carbonate group reacts with amines for versatile payload attachment.
  • High purity (95.0%), white to off-white solid.
  • Soluble in DMSO (100 mg/mL; may require ultrasonic treatment).
  • Storage: powder -20 °C; in solvent -80 °C (6 months) or -20 °C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.