Learn More
Hach Company PC/RC Combo Digital Gateway
Supplier:  Hach Company 6120600
Encompass
PC/RC Combination Digital GatewayDigital Gateway to convert PC/RC analog pH sensors to digital output for connecting to sc digitial controller
Catalog No. 50-002-07874