Learn More
Medchemexpress LLC PC SPDP-NHS carbonate ester | 2279944-61-3 | 95.0% | 608.64 g/mol | C25H28N4O10S2 | 1 MG
SDP
Catalog No. 5000203779
Encompass_Preferred
PC SPDP-NHS carbonate ester is a cleavable linker used in the synthesis of antibody-drug conjugates (ADCs); it contains an NHS carbonate for amine conjugation and a photolabile SPDP moiety that enables controlled release of payloads.
  • Cleavable linker suitable for ADC synthesis.
  • Contains NHS carbonate for amine conjugation.
  • Photolabile SPDP moiety enables controlled release.
  • High purity appropriate for research use.
  • Supplied in milligram-scale quantities for laboratory applications.
Catalog No. 50-002-03779 Supplier Medchemexpress LLC Supplier No. HY1401381MG
PC SPDP-NHS carbonate ester is a cleavable linker used in the synthesis of antibody-drug conjugates (ADCs); it contains an NHS carbonate for amine conjugation and a photolabile SPDP moiety that enables controlled release of payloads.
  • Cleavable linker suitable for ADC synthesis.
  • Contains NHS carbonate for amine conjugation.
  • Photolabile SPDP moiety enables controlled release.
  • High purity appropriate for research use.
  • Supplied in milligram-scale quantities for laboratory applications.