Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PC SPDP-NHS carbonate ester | 2279944-61-3 | MFCD31811480 | 95.0% | 608.64 | C25H28N4O10S2 | 5 MG
SDP

Catalog No. 5000203780
Encompass_Preferred

PC SPDP-NHS carbonate ester is a photocleavable carbonate NHS ester linker used for bioconjugation and the construction of antibody-drug conjugates (ADCs). It provides an NHS carbonate reactive site for coupling to primary amines and a photolabile unit that enables controlled payload release.

  • Cleavable photocleavable linker suitable for ADC synthesis
  • Contains an NHS carbonate reactive group for amine conjugation
  • Purity 95.0%
  • Molecular weight 608.64
  • Soluble in common organic solvents such as DMSO
  • Supplied in small research pack sizes for laboratory use

Catalog No. 50-002-03780 Supplier Medchemexpress LLC Supplier No. HY1401385MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PC SPDP-NHS carbonate ester is a photocleavable carbonate NHS ester linker used for bioconjugation and the construction of antibody-drug conjugates (ADCs). It provides an NHS carbonate reactive site for coupling to primary amines and a photolabile unit that enables controlled payload release.

  • Cleavable photocleavable linker suitable for ADC synthesis
  • Contains an NHS carbonate reactive group for amine conjugation
  • Purity 95.0%
  • Molecular weight 608.64
  • Soluble in common organic solvents such as DMSO
  • Supplied in small research pack sizes for laboratory use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.