Learn More
World Precision Instrument PC to pump cable, 7 ft
SDP
Catalog No. 502538794
Encompass
The Aladdin series of syringe pumps provides the capability to both inject and withdraw. These pumps can be used for manually triggered injections, but also have the capability to be programmed in multi-step pumping sequences without the requirement for a computer. Even so, this series also has both RS-232 and TTL inputs for external control and status feedback to a computer, if desired. The pumps can be daisy chained for multi pump applications involving push/pull protocols or multiple independent channels.
This cable connects your Aladdin pump to a computer.
Catalog No. 50-253-8794 Supplier World Precision Instrument Supplier No. GNPC7
The Aladdin series of syringe pumps provides the capability to both inject and withdraw. These pumps can be used for manually triggered injections, but also have the capability to be programmed in multi-step pumping sequences without the requirement for a computer. Even so, this series also has both RS-232 and TTL inputs for external control and status feedback to a computer, if desired. The pumps can be daisy chained for multi pump applications involving push/pull protocols or multiple independent channels.
This cable connects your Aladdin pump to a computer.