Learn More
Enzo Life Sciences PC12 (heat shocked) , (cell lysate) (200µg)
SDP
Supplier:  Enzo Life Sciences ADILYCPC101F
Encompass_Preferred
Formulation: Liquid. In SDS-PAGE sample buffer, pH 6.8, heated for 10 min. at 70°C and containing 40mM TRIS-HCl, 1% SDS, 50mM DTT, 7.5% glycerol, and 0.003% bromophenol blue. Source: Rat. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles. Applications: WB .
Catalog No. 50-200-7337