Learn More
Cole-Parmer Environmental Express Cole Parmer PC200-02 pH/Conductivity Meter Kit; Cell, 3-in-1 pH Electrode, Holder
Catalog No. NC1890417 Shop All Cole-Parmer Products
Encompass_Preferred
The P200 series of benchtop meters provides accurate, reliable results with the benefit of convenient accessories to make your lab run smoothly. Large color TFT display is easily visible, and features a stability icon to alert you when your readings are ready to be recorded and an electrode health icon indicating when you need to refill or replace your electrode.
Catalog No. NC1890417 Supplier Cole-Parmer Supplier No. 5925015
The P200 series of benchtop meters provides accurate, reliable results with the benefit of convenient accessories to make your lab run smoothly. Large color TFT display is easily visible, and features a stability icon to alert you when your readings are ready to be recorded and an electrode health icon indicating when you need to refill or replace your electrode.