Learn More
Perkin Elmer US LLC PC3 Peltier Heated and Cooled Cyclonic Spray Chamber for Avio 200
Catalog No. 502105387
Encompass_Preferred
The PC3x for Avio is a compact, Peltier-heated or cooled inlet system which incorporates the ESI cyclonic spray chamber. In cooling mode the PC3x cools the outer walls of the cyclonic spray chamber that operates at -10°C to +80°C, reducing the water or solvent vapor loading on the plasma, resulting in enhanced stability and performance. In heating mode, sensitivity can be improved for low flow rate applications.
Catalog No. 50-210-5387 Supplier Perkin Elmer US LLC Supplier No. N0790959
Add to Cart
Add to Cart
Request a Quote
The PC3x for Avio is a compact, Peltier-heated or cooled inlet system which incorporates the ESI cyclonic spray chamber. In cooling mode the PC3x cools the outer walls of the cyclonic spray chamber that operates at -10°C to +80°C, reducing the water or solvent vapor loading on the plasma, resulting in enhanced stability and performance. In heating mode, sensitivity can be improved for low flow rate applications.