Learn More
ATCC PC-3; Prostate Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502384682
Encompass
PC-3 is a cell line initiated from a bone metastasis of a grade IV prostatic adenocarcinoma from a 62-year-old, White, male.
Catalog No. 50-238-4682 Supplier ATCC Supplier No. CRL1435
PC-3 is a cell line initiated from a bone metastasis of a grade IV prostatic adenocarcinoma from a 62-year-old, White, male.