Learn More
Perkin Elmer US LLC PC3x Peltier Cooled Cyclonic Spray Chamber for Avio 500
Catalog No. 502113258
Encompass_Preferred
Benefits and features: Temperature control from -10 °C to +80 °C Controlled directly through Syngistix Thermal stabilization of spray chamber improves long-term stability Last set temperature saved for stand-alone operation Remote monitoring and control The PC3Xfor cyclonic spray chambers is included in the NexION 2000 ICP-MS Core Configuration's Sample Introduction Kit (N8150035), the Productivity Configuration's Sample Introduction Kit (N8150036), and the Semi Configuration's Sample Introduction Kit (N8150037).
Catalog No. 50-211-3258 Supplier Perkin Elmer US LLC Supplier No. N0810860
Add to Cart
Add to Cart
Request a Quote
Benefits and features: Temperature control from -10 °C to +80 °C Controlled directly through Syngistix Thermal stabilization of spray chamber improves long-term stability Last set temperature saved for stand-alone operation Remote monitoring and control The PC3Xfor cyclonic spray chambers is included in the NexION 2000 ICP-MS Core Configuration's Sample Introduction Kit (N8150035), the Productivity Configuration's Sample Introduction Kit (N8150036), and the Semi Configuration's Sample Introduction Kit (N8150037).