Learn More
Perkin Elmer US LLC PC3X Peltier Cooler for PFA Barrel Spray Chamber
Catalog No. 502113298
Encompass_Preferred
Features and benefits: Variable temperature control from -10 °C to +80 °C Thermal stabilization of spray chamber improves long-term stability USB or Bluetooth connectivity Remote monitoring and control
Catalog No. 50-211-3298 Supplier Perkin Elmer US LLC Supplier No. N8150451
Add to Cart
Add to Cart
Request a Quote
Features and benefits: Variable temperature control from -10 °C to +80 °C Thermal stabilization of spray chamber improves long-term stability USB or Bluetooth connectivity Remote monitoring and control