Learn More
Perkin Elmer US LLC PC3x Peltier Cooler without Spray Chamber for NexION 350
Catalog No. 502104110
Encompass_Preferred
The PC3x is a compact, Peltier heated or cooled inlet system that controls the spray chamber temperature and improves long-term stability. In cooling mode, water or solvent is removed from the aerosol, reducing water or solvent loading on the plasma. In heating mode, sensitivity is increased and volatility-related sample transport effects can be reduced or eliminated. Variable temperature control from -10 °C to +80 °C Thermal stabilization of spray chamber improves long-term stability Last set temperature saved for stand alone operation USB or Bluetooth connectivity Remote monitoring and control
Catalog No. 50-210-4110 Supplier Perkin Elmer US LLC Supplier No. N8140672
The PC3x is a compact, Peltier heated or cooled inlet system that controls the spray chamber temperature and improves long-term stability. In cooling mode, water or solvent is removed from the aerosol, reducing water or solvent loading on the plasma. In heating mode, sensitivity is increased and volatility-related sample transport effects can be reduced or eliminated. Variable temperature control from -10 °C to +80 °C Thermal stabilization of spray chamber improves long-term stability Last set temperature saved for stand alone operation USB or Bluetooth connectivity Remote monitoring and control