Learn More
Perkin Elmer US LLC PC3X Peltier Heater/Cooler Sample Introduction Kit for Optima 2x00/3x00/4x00/5x00/7x00 DV/8x00
Catalog No. 502104118
Encompass_Preferred
The PC3X is a compact, Peltier-heated or cooled inlet system which incorporates the ESI cyclonic spray chamber. In cooling mode the PC3X heats or cools the outer walls of the cyclonic spray chamber, reducing the water or solvent vapor loading on the plasma, resulting in enhanced stability and performance. In heating mode, sensitivity can be improved for low flow rate applications. Temperature control from -10 °C to +80 °C Thermal stabilization of spray chamber improves long-term stability Last set temperature saved for stand-alone operation USB or Bluetooth connectivity Remote monitoring and control This sample introduction kit includes: PC3X Peltier Heater/Cooler and Power Supply Baffled Quartz Cyclonic Spray Chamber PFA-ST MicroFlow Nebulizer (N0777049) 1.5 mm I. D. Quartz Convertible Injector 0.15 mm I.D. PFA Sampling Tubing, 1 m (N8122382) 0.25 mm I.D. PFA Sampling Tubing, 1 m (N8122383) 0.5 mm I.D. PFA Sampling Tubing, 1 m (N8122384) Tee Connector for Mixing Oxygen and Make-Up G
Catalog No. 50-210-4118 Supplier Perkin Elmer US LLC Supplier No. N0777836
Add to Cart
Add to Cart
Request a Quote
The PC3X is a compact, Peltier-heated or cooled inlet system which incorporates the ESI cyclonic spray chamber. In cooling mode the PC3X heats or cools the outer walls of the cyclonic spray chamber, reducing the water or solvent vapor loading on the plasma, resulting in enhanced stability and performance. In heating mode, sensitivity can be improved for low flow rate applications. Temperature control from -10 °C to +80 °C Thermal stabilization of spray chamber improves long-term stability Last set temperature saved for stand-alone operation USB or Bluetooth connectivity Remote monitoring and control This sample introduction kit includes: PC3X Peltier Heater/Cooler and Power Supply Baffled Quartz Cyclonic Spray Chamber PFA-ST MicroFlow Nebulizer (N0777049) 1.5 mm I. D. Quartz Convertible Injector 0.15 mm I.D. PFA Sampling Tubing, 1 m (N8122382) 0.25 mm I.D. PFA Sampling Tubing, 1 m (N8122383) 0.5 mm I.D. PFA Sampling Tubing, 1 m (N8122384) Tee Connector for Mixing Oxygen and Make-Up G