Learn More
Perkin Elmer US LLC PC3X Spray Chamber/Cassette UV Light Shield for NexION 1000/2000
Catalog No. 502103967
Encompass_Preferred
The UV light shield protects users from rays given off by the plasma in the instrument.
Catalog No. 50-210-3967 Supplier Perkin Elmer US LLC Supplier No. N8152475
The UV light shield protects users from rays given off by the plasma in the instrument.