Learn More
Perkin Elmer US LLC PC3X Thermally Stabilized Inlet System with PFA Spray Chamber for NexION 1000/2000
Catalog No. 502103740
Encompass_Preferred
Assembly includes: Peltier Cooler/Heater Thermally Stabilized Inlet System (N8150451) Mounting Post (N8152723) High Performance PFA Spray Chamber (N8150454) Metal Free Addition Gas Line, 0.8 mm I.D., 10 in. (N8152449) Spray chamber Drain Line (N8152456) Note: Does not include PFA Nebulizer. PC3x Thermally Stabilized Inlet System with Nebulizer and Spray Chamber (N8150446) available separately.
Catalog No. 50-210-3740 Supplier Perkin Elmer US LLC Supplier No. N8150445
Add to Cart
Add to Cart
Request a Quote
Assembly includes: Peltier Cooler/Heater Thermally Stabilized Inlet System (N8150451) Mounting Post (N8152723) High Performance PFA Spray Chamber (N8150454) Metal Free Addition Gas Line, 0.8 mm I.D., 10 in. (N8152449) Spray chamber Drain Line (N8152456) Note: Does not include PFA Nebulizer. PC3x Thermally Stabilized Inlet System with Nebulizer and Spray Chamber (N8150446) available separately.