Learn More
Aviva Systems Biology Corp PC4 Antibody - N-terminal region (P100959_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100959_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PC4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PC4. Alternate names: P15, PC4, p14ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Rat
Catalog No. 50-014-62801