Learn More
Electron Microscopy Sciences PC620 Hot Plate/Stirrer, 120 V, 60-1150 RPM
SDP
Catalog No. 501899837
Encompass_Preferred
Pyroceram glass-ceramic stirring hot plates features a microprocessor with digital display that regulates heating and stirring. Consistent and repeatable temperature is maintained from 25 °C up to 550 °C and is adjustable in 5 °C increments. Display blinks until temp is reached and LED HOT TOP icon lights up when top plate is over 60°C and separate temp sensor provides power cutoff if unit overheats.An optional external temp controller is available for inside the vessel and will shut off unit when is not in liquid. Stirring speed range is from 60-1150 RPM. Stronger, larger magnet improves stir bar coupling combined with automatic up speed adjustment minimizes decoupling. The two piece casting design deflects spills away from electronic and includes a built in mount for an optional 18" support rod. UL and cUL approved.Heating surface: 10" x 10"Range: 25-550°CRPM: 60-1150Weight: 11.5 lbFootprint: 15.8 x 10.6
Catalog No. 50-189-9837 Supplier Electron Microscopy Sciences Supplier No. 7234910
Pyroceram glass-ceramic stirring hot plates features a microprocessor with digital display that regulates heating and stirring. Consistent and repeatable temperature is maintained from 25 °C up to 550 °C and is adjustable in 5 °C increments. Display blinks until temp is reached and LED HOT TOP icon lights up when top plate is over 60°C and separate temp sensor provides power cutoff if unit overheats.An optional external temp controller is available for inside the vessel and will shut off unit when is not in liquid. Stirring speed range is from 60-1150 RPM. Stronger, larger magnet improves stir bar coupling combined with automatic up speed adjustment minimizes decoupling. The two piece casting design deflects spills away from electronic and includes a built in mount for an optional 18" support rod. UL and cUL approved.Heating surface: 10" x 10"Range: 25-550°CRPM: 60-1150Weight: 11.5 lbFootprint: 15.8 x 10.6