Learn More
MilliporeSigma Milliflex Cassette prefilled with Plate Count Agar, 10ml
Catalog No. NC9481836
Encompass
Milliflex Cassette prefilled with Plate Count Agar. This medium is designed for the recovery of bacteria in water, waste water and dairy products (aerobic microorganisms), 10ml
Catalog No. NC9481836 Supplier MilliporeSigma Supplier No. MXSMPCA48
Milliflex Cassette prefilled with Plate Count Agar. This medium is designed for the recovery of bacteria in water, waste water and dairy products (aerobic microorganisms), 10ml