Learn More
Aviva Systems Biology Corp PCBD2 Antibody - N-terminal region (ARP60485_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60485_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCBD2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PCBD2. Alternate names: DCOH2, DCOHM, PHS2ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-287-061