Learn More
Aviva Systems Biology Corp PCBD2 Antibody - C-terminal region (ARP60484_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60484_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCBD2. It was validated on Western Blot. . Alternate names: DCOH2, DCOHM, PHS2ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-287-060