Learn More
Aviva Systems Biology Corp PCBP1 Antibody - middle region (ARP40630_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40630_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PCBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PCBP1. Alternate names: HNRPX, HNRPE1, hnRNP-X, hnRNP-E1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57558