Learn More
Molecular Devices Molecular Devices Wash Head, 96-Well, AquaMax 2000, AquaMax 4000
Catalog No. 5000014724
Encompass_Preferred
The 96-Well Microplate Wash Head (0310-5216T) is a replacement wash head component manufactured for use with Molecular Devices AquaMax 2000 and AquaMax 4000 microplate washers. It provides standard 96-well plate washing capability and installs into the washer's wash head assembly.
  • Supports 96-well microplate format.
  • Provides uniform well washing across standard microplates.
  • Designed for straightforward installation into the wash head assembly.
  • Suitable for routine plate washing and cell-wash protocols.
  • Supplied as a component for wash head maintenance and replacement.
Catalog No. 50-000-14724 Supplier Molecular Devices Supplier No. 03105216T
The 96-Well Microplate Wash Head (0310-5216T) is a replacement wash head component manufactured for use with Molecular Devices AquaMax 2000 and AquaMax 4000 microplate washers. It provides standard 96-well plate washing capability and installs into the washer's wash head assembly.
  • Supports 96-well microplate format.
  • Provides uniform well washing across standard microplates.
  • Designed for straightforward installation into the wash head assembly.
  • Suitable for routine plate washing and cell-wash protocols.
  • Supplied as a component for wash head maintenance and replacement.