Learn More
System Biosciences LLC pCDH-CMV-MCS-EF1α-RFP+Puro Cloning and Expression LentivectorpCDH-CMV-MCS-EF1α-RFP+Puro cDNA Dual Promoter Cloning and Expression Lentivector = 10ug
SDP
Supplier:  System Biosciences LLC CD516B2
Encompass_Preferred
With the strong CMV promoter upstream of an MCS and EF1α driving an RFP-puromycin resistance fusion, pCDH-CMV-MCS-EF1α-RFP+Puro efficiently delivers two markers. Well-regarded in the industry for high, reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-CMV-MCS-EF1α-RFP+Puro Cloning and Expression Lentivector drives expression of your gene-of-interest from the strong CMV promoter, and expression of an RFP-puromycin resistance fusion from the EF1α promoter.
Catalog No. NC1169792