Learn More
System Biosciences LLC pCDH-CMV-MCS-EF1α-GreenPuro Cloning and Expression LentivectorpCDH-CMV-MCS-EF1α-GreenPuro cDNA Dual Promoter Cloning and Expression Lentivector = 10ug
SDP
Catalog No. NC1395454
Encompass_Preferred
With the strong CMV promoter upstream of an MCS and EF1α driving a GFP-puromycin resistance fusion, pCDH-CMV-MCS-EF1α-GreenPuro Lentivector efficiently provides two markers. Well-regarded in the industry for high, reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-CMV-MCS-EF1α-GreenPuro Cloning and Expression Lentivector drives expression of your gene-of-interest from the strong CMV promoter, and expression of a GFP-puromycin resistance fusion gene from the EF1α promoter.
Catalog No. NC1395454 Supplier System Biosciences LLC Supplier No. CD513B1
With the strong CMV promoter upstream of an MCS and EF1α driving a GFP-puromycin resistance fusion, pCDH-CMV-MCS-EF1α-GreenPuro Lentivector efficiently provides two markers. Well-regarded in the industry for high, reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-CMV-MCS-EF1α-GreenPuro Cloning and Expression Lentivector drives expression of your gene-of-interest from the strong CMV promoter, and expression of a GFP-puromycin resistance fusion gene from the EF1α promoter.