Learn More
System Biosciences LLC pCDH-CMV-MCS-EF1α-Puro Cloning and Expression LentivectorpCDH-CMV-MCS-EF1α-Puro cDNA Dual Promoter Cloning and Expression Lentivector = 10ug
SDP
Catalog No. NC1458360
Encompass_Preferred
With the strong CMV promoter upstream of an MCS and EF1α driving the puromycin resistance marker, pCDH-CMV-MCS-EF1α-Puro is a great, all-purpose lentivector. Well-regarded in the industry for high, reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-CMV-MCS-EF1α-Puro Cloning and Expression Lentivector drives expression of your gene-of-interest from the strong CMV promoter, and expression of the puromycin resistance marker from the EF1α promoter.
Catalog No. NC1458360 Supplier System Biosciences LLC Supplier No. CD510B1
With the strong CMV promoter upstream of an MCS and EF1α driving the puromycin resistance marker, pCDH-CMV-MCS-EF1α-Puro is a great, all-purpose lentivector. Well-regarded in the industry for high, reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-CMV-MCS-EF1α-Puro Cloning and Expression Lentivector drives expression of your gene-of-interest from the strong CMV promoter, and expression of the puromycin resistance marker from the EF1α promoter.