Learn More
System Biosciences LLC pCDH-CMV-MCS-EF1α-RFP Cloning and Expression LentivectorpCDH-CMV-MCS-EF1α-RFP cDNA Dual Promoter Cloning and Expression Lentivector = >1E6 IFUs
SDP
Catalog No. NC2258575
Encompass_Preferred
With the strong CMV promoter upstream of an MCS and EF1α driving the RFP reporter, pCDH-CMV-MCS-EF1α-RFP is a great, all-purpose lentivector. Well-regarded in the industry for high, reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-CMV-MCS-EF1α-RFP Cloning and Expression Lentivector drives expression of your gene-of-interest from the strong CMV promoter, and expression of the RFP reporter from the EF1α promoter.
This item is not returnable. View return policy
Catalog No. NC2258575 Supplier System Biosciences LLC Supplier No. CD512B1
With the strong CMV promoter upstream of an MCS and EF1α driving the RFP reporter, pCDH-CMV-MCS-EF1α-RFP is a great, all-purpose lentivector. Well-regarded in the industry for high, reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-CMV-MCS-EF1α-RFP Cloning and Expression Lentivector drives expression of your gene-of-interest from the strong CMV promoter, and expression of the RFP reporter from the EF1α promoter.