Learn More
System Biosciences LLC pCDH-CuO-MCS-EF1α-CymR-T2A-Puro SparQ™ All-in-one Cloning and Expression LentivectorpCDH-Cuo-MCS-EF1α-CymR-T2A-Puro = 10 µg
SDP
Catalog No. NC1820538
Encompass_Preferred
Simplify your SparQ projects by delivering your gene-of-interest and CymR with the same lentivector – includes co-expression of CymR and a puromycin marker. With SBI’s SparQ™ Cumate Switch System, you can get inducible gene expression in mammalian cells with a range of cloning and expression lentivectors. The pCDH-CuO-MCS-EF1α-CymR-T2A-Puro SparQ All-in-one Cloning and Expression Lentivector (Cat.# QM800A-1) simplifies your SparQ projects by delivering your gene- or miRNA-of-interest with the same vector that delivers CymR. The vector drives your gene-of-interest or miRNA-of-interest with the inducible cumate switch promoter, and constitutively co-expresses CymR and a puromycin resistance marker from an EF1α promoter. Co-expression is mediated by the T2A element.
Catalog No. NC1820538 Supplier System Biosciences LLC Supplier No. QM800A1
Simplify your SparQ projects by delivering your gene-of-interest and CymR with the same lentivector – includes co-expression of CymR and a puromycin marker. With SBI’s SparQ™ Cumate Switch System, you can get inducible gene expression in mammalian cells with a range of cloning and expression lentivectors. The pCDH-CuO-MCS-EF1α-CymR-T2A-Puro SparQ All-in-one Cloning and Expression Lentivector (Cat.# QM800A-1) simplifies your SparQ projects by delivering your gene- or miRNA-of-interest with the same vector that delivers CymR. The vector drives your gene-of-interest or miRNA-of-interest with the inducible cumate switch promoter, and constitutively co-expresses CymR and a puromycin resistance marker from an EF1α promoter. Co-expression is mediated by the T2A element.