Learn More
System Biosciences LLC pCDH-EF1α-MCS-IRES-Puro Cloning and Expression LentivectorpCDH-EF1α-MCS-IRES-Puro cDNA Single Promoter Cloning and Expression Lentivector = 10ug
SDP
Catalog No. NC2011214
Encompass_Preferred
With the moderate EF1α promoter upstream of an MCS and an IRES element to co-express a Puro marker, this lentivector is great for stable gene expression. Well-regarded in the industry for high, reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-EF1α-MCS-IRES-Puro Cloning and Expression Lentivector uses the moderate EF1α promoter to co-express your gene of interest with a puromycin resistance marker, thanks to a IRES element.
Catalog No. NC2011214 Supplier System Biosciences LLC Supplier No. CD532A2
With the moderate EF1α promoter upstream of an MCS and an IRES element to co-express a Puro marker, this lentivector is great for stable gene expression. Well-regarded in the industry for high, reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-EF1α-MCS-IRES-Puro Cloning and Expression Lentivector uses the moderate EF1α promoter to co-express your gene of interest with a puromycin resistance marker, thanks to a IRES element.