Learn More
System Biosciences LLC pCDH-EF1α-MCS-(PGK-GFP) Bidirectional Promoter Cloning and Expression LentivectorpCDH-EF1α-MCS-(PGK-GFP) Bidirectional Promoter, Cloning and Expression Lentivector = 10ug
SDP
Catalog No. NC1669739
Encompass_Preferred
Get another option for robust, coordinated dual gene expression with the pCDH-EF1α-MCS-(PGK-GFP) Bidirectional Promoter Lentivector. Well-regarded in the industry for reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-EF1α-MCS-(PGK-GFP) Bidirectional Promoter Cloning and Expression Lentivector gives you another option for robust, coordinated expression of your gene-of-interest and a GFP reporter.
Catalog No. NC1669739 Supplier System Biosciences LLC Supplier No. CD811A1
Get another option for robust, coordinated dual gene expression with the pCDH-EF1α-MCS-(PGK-GFP) Bidirectional Promoter Lentivector. Well-regarded in the industry for reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-EF1α-MCS-(PGK-GFP) Bidirectional Promoter Cloning and Expression Lentivector gives you another option for robust, coordinated expression of your gene-of-interest and a GFP reporter.