Learn More
System Biosciences LLC pCDH-EF1α-MCS-(PGK-GFP-T2A-Puro) Bidirectional Promoter Cloning and Expression LentivectorpCDH-EF1α-MCS-(PGK-GFP-T2A-Puro) Cloning and Expression Lentivector = 10ug
SDP
Supplier:  System Biosciences LLC CD813A1
Encompass_Preferred
Get another option for robust, coordinated dual gene expression with the pCDH-EF1α-MCS-(PGK-GFP-T2A-Puro) Bidirectional Promoter Lentivector. Well-regarded in the industry for reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-EF1α-MCS-(PGK-GFP-T2A-Puro) Bidirectional Promoter Cloning and Expression Lentivector gives you another option for robust, coordinated expression of your gene-of-interest and co-expressed GFP and puromycin resistance via a T2A element.
Catalog No. NC1763699