Learn More
System Biosciences LLC pCDH-MSCV-MCS-EF1α-GFP Cloning and Expression LentivectorpCDH-MSCV-MCS-EF1α-GFP cDNA Dual Promoter Cloning and Expression Lentivector = 10ug
SDP
Catalog No. NC2258577
Encompass_Preferred
The pCDH-MSCV-MCS-EF1α-GFP lentivector uses the MSCV promoter for high expression in hematopoietic or stem cells and includes a GFP reporter. Well-regarded in the industry for reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-MSCV-MCS-EF1α-GFP Cloning and Expression Lentivector drives expression of your gene-of-interest from the MSCV promoter, which provides strong expression in hematopoietic and stem cells. This lentivector also enables selection for transductants using a GFP reporter.
This item is not returnable. View return policy
Catalog No. NC2258577 Supplier System Biosciences LLC Supplier No. CD711B1
The pCDH-MSCV-MCS-EF1α-GFP lentivector uses the MSCV promoter for high expression in hematopoietic or stem cells and includes a GFP reporter. Well-regarded in the industry for reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-MSCV-MCS-EF1α-GFP Cloning and Expression Lentivector drives expression of your gene-of-interest from the MSCV promoter, which provides strong expression in hematopoietic and stem cells. This lentivector also enables selection for transductants using a GFP reporter.